Merck
  • Tenascin-C Orchestrates an Immune-Suppressive Tumor Microenvironment in Oral Squamous Cell Carcinoma.
Tenascin-C Orchestrates an Immune-Suppressive Tumor Microenvironment in Oral Squamous Cell Carcinoma.
Cancer immunology research (2020-07-16)
Caroline Spenlé, Thomas Loustau, Devadarssen Murdamoothoo, William Erne, Stephanie Beghelli-de la Forest Divonne, Romain Veber, Luciana Petti, Pierre Bourdely, Matthias Mörgelin, Eva-Maria Brauchle, Gérard Cremel, Vony Randrianarisoa, Abdouramane Camara, Samah Rekima, Sebastian Schaub, Kelly Nouhen, Thomas Imhof, Uwe Hansen, Nicodème Paul, Raphael Carapito, Nicolas Pythoud, Aurélie Hirschler, Christine Carapito, Hélène Dumortier, Christopher G Mueller, Manuel Koch, Katja Schenke-Layland, Shigeyuki Kon, Anne Sudaka, Fabienne Anjuère, Ellen Van Obberghen-Schilling, Gertraud Orend
ABSTRACT
Inherent immune suppression represents a major challenge in the treatment of human cancer. The extracellular matrix molecule tenascin-C promotes cancer by multiple mechanisms, yet the roles of tenascin-C in tumor immunity are incompletely understood. Using a 4NQO-induced oral squamous cell carcinoma (OSCC) model with abundant and absent tenascin-C, we demonstrated that tenascin-C enforced an immune-suppressive lymphoid stroma via CCL21/CCR7 signaling, leading to increased metastatic tumors. Through TLR4, tenascin-C increased expression of CCR7 in CD11c+ myeloid cells. By inducing CCL21 in lymphatic endothelial cells via integrin α9β1 and binding to CCL21, tenascin-C immobilized CD11c+ cells in the stroma. Inversion of the lymph node-to-tumor CCL21 gradient, recruitment of T regulatory cells, high expression of anti-inflammatory cytokines, and matrisomal components were hallmarks of the tenascin-C-instructed lymphoid stroma. Ablation of tenascin-C or CCR7 blockade inhibited the lymphoid immune-suppressive stromal properties, reducing tumor growth, progression, and metastasis. Thus, targeting CCR7 could be relevant in human head and neck tumors, as high tenascin-C expression and an immune-suppressive stroma correlate to poor patient survival.
Sigma-Aldrich
MISSION® shRNA, Lentiviral Particles
Sigma-Aldrich
Eosin Y Solution, Alcoholic
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.