Merck
  • MFSD7C switches mitochondrial ATP synthesis to thermogenesis in response to heme.
MFSD7C switches mitochondrial ATP synthesis to thermogenesis in response to heme.
Nature communications (2020-09-26)
Yingzhong Li, Nikola A Ivica, Ting Dong, Dimitrios P Papageorgiou, Yanpu He, Douglas R Brown, Marianna Kleyman, Guangan Hu, Walter W Chen, Lucas B Sullivan, Amanda Del Rosario, Paula T Hammond, Matthew G Vander Heiden, Jianzhu Chen
ABSTRACT
ATP synthesis and thermogenesis are two critical outputs of mitochondrial respiration. How these outputs are regulated to balance the cellular requirement for energy and heat is largely unknown. Here we show that major facilitator superfamily domain containing 7C (MFSD7C) uncouples mitochondrial respiration to switch ATP synthesis to thermogenesis in response to heme. When heme levels are low, MSFD7C promotes ATP synthesis by interacting with components of the electron transport chain (ETC) complexes III, IV, and V, and destabilizing sarcoendoplasmic reticulum Ca2+-ATPase 2b (SERCA2b). Upon heme binding to the N-terminal domain, MFSD7C dissociates from ETC components and SERCA2b, resulting in SERCA2b stabilization and thermogenesis. The heme-regulated switch between ATP synthesis and thermogenesis enables cells to match outputs of mitochondrial respiration to their metabolic state and nutrient supply, and represents a cell intrinsic mechanism to regulate mitochondrial energy metabolism.
Sigma-Aldrich
N-Ethylacrylamide, contains MEHQ as inhibitor, 99% (HPLC)
Sigma-Aldrich
ADP Assay Kit, sufficient for 100 assays (bioluminescent)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Anti-FLVCR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution