Merck
  • A system-level approach identifies HIF-2α as a critical regulator of chondrosarcoma progression.
A system-level approach identifies HIF-2α as a critical regulator of chondrosarcoma progression.
Nature communications (2020-10-08)
Hyeonkyeong Kim, Yongsik Cho, Hyeon-Seop Kim, Donghyun Kang, Donghyeon Cheon, Yi-Jun Kim, Moon Jong Chang, Kyoung Min Lee, Chong Bum Chang, Seung-Baik Kang, Hyun Guy Kang, Jin-Hong Kim
ABSTRACT
Chondrosarcomas, malignant cartilaginous neoplasms, are capable of transitioning to highly aggressive, metastatic, and treatment-refractory states, resulting in significant patient mortality. Here, we aim to uncover the transcriptional program directing such tumor progression in chondrosarcomas. We conduct weighted correlation network analysis to extract a characteristic gene module underlying chondrosarcoma malignancy. Hypoxia-inducible factor-2α (HIF-2α, encoded by EPAS1) is identified as an upstream regulator that governs the malignancy gene module. HIF-2α is upregulated in high-grade chondrosarcoma biopsies and EPAS1 gene amplification is associated with poor prognosis in chondrosarcoma patients. Using tumor xenograft mouse models, we demonstrate that HIF-2α confers chondrosarcomas the capacities required for tumor growth, local invasion, and metastasis. Meanwhile, pharmacological inhibition of HIF-2α, in conjunction with the chemotherapy agents, synergistically enhances chondrosarcoma cell apoptosis and abolishes malignant signatures of chondrosarcoma in mice. We expect that our insights into the pathogenesis of chondrosarcoma will provide guidelines for the development of molecular targeted therapeutics for chondrosarcoma.
Sigma-Aldrich
Annexin V-FITC Apoptosis Detection Kit
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse