Merck
  • Non-targeted screening of plastic-related chemicals in food collected in Montreal, Canada.
Non-targeted screening of plastic-related chemicals in food collected in Montreal, Canada.
Food chemistry (2020-05-15)
Lei Tian, Jingyun Zheng, Cindy Gates Goodyer, Stéphane Bayen
ABSTRACT
A non-targeted screening method based on ultrasound-assisted extraction followed by high-performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry (HPLC-QTOF-MS) was developed to screen for the presence of plastic-related chemicals (PRCs) in different types of food (fish, chicken, canned tuna, leafy vegetables, bread and butter). Eleven bisphenols were used as targeted compounds. Instrument linearity (r2 ≥0.98), inter-day precision (RSD ≤9.0%) as well as method detection limits (MDLs below 3.6 ng g-1) were satisfactory. Recoveries of the eleven bisphenols ranged from 76% to 122% among the different food matrices. The method was applied to food collected from Montreal, Canada in 2017-2018. The non-targeted screening approach identified a range of contaminants in different food matrices, including BPA, BPS, bis(2-ethylhexyl) adipate, dibutyl adipate, hexadecyl methacrylate and Irganox®1076. Further research is suggested to investigate the concentration of these PRCs, the consumption habits of average and specific populations and the potential routes of contamination.
Sigma-Aldrich
Dibutyl adipate, 96%
Sigma-Aldrich
4,4′-Cyclohexylidenebisphenol, 98%
Sigma-Aldrich
4,4′-(1-Phenylethylidene)bisphenol, 99%