Merck
  • rec-Y3H screening allows the detection of simultaneous RNA-protein interface mutations.
rec-Y3H screening allows the detection of simultaneous RNA-protein interface mutations.
Methods (San Diego, Calif.) (2019-09-08)
Mireia Garriga-Canut, Jae-Seong Yang, Friedrich Preusser, Silvia Speroni, Maria Gili, Sebastian P Maurer
ABSTRACT
Understanding which proteins and RNAs directly interact is crucial for revealing cellular mechanisms of gene regulation. Efficient methods allowing to detect RNA-protein interactions and dissect the underlying molecular origin for RNA-binding protein (RBP) specificity are in high demand. The recently developed recombination-Y3H screening (rec-Y3H) enabled many-by-many detection of interactions between pools of proteins and RNA fragments for the first time. Here, we test different conditions for protein-RNA interaction selection during rec-Y3H screening and provide information on the screen performance in several selection media. We further show that rec-Y3H can detect the nucleotide and amino acid sequence determinants of protein-RNA interactions by mutating residues of interacting proteins and RNAs simultaneously. We envision that systematic RNA-protein interface mutation screening will be useful to understand the molecular origin of RBP selectivity and to engineer RBPs with targeted specificities in the future.
Sigma-Aldrich
Poly(ethylene glycol), BioXtra, average mol wt 3,350, powder
Sigma-Aldrich
Adenine hemisulfate salt, ≥99%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Lithium acetate dihydrate, BioXtra
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Amino-1,2,4-triazole, ≥95% (TLC)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Sigma-Aldrich
Lithium acetate, 99.95% trace metals basis
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra