Merck
  • Chemokine CXCL14-like immunoreactivity in the αMSH-producing cells and PRL-producing cells of the flat-tailed house gecko pituitary.
Chemokine CXCL14-like immunoreactivity in the αMSH-producing cells and PRL-producing cells of the flat-tailed house gecko pituitary.
The Journal of veterinary medical science (2020-02-11)
Hirohumi Suzuki, Toshiharu Yamamoto
ABSTRACT
The distribution pattern of chemokine CXCL14-immunoreactive cells was examined by immunohistochemistry in the pituitary of the gecko Hemidactylus platyurus. Immunoreactive cells were observed in the pars intermedia and pars distalis of the pituitary, but not in the pars nervosa. All α-melanocyte-stimulating hormone (αMSH)-producing cells were immunoreactive for CXCL14 in the pars intermedia. The CXCL14-immunoreactive cells corresponded to prolactin (PRL)-producing cells but not to other adenohypophyseal-hormone-producing cells in the pars distalis. CXCL14 secreted from αMSH-producing cells and PRL-producing cells may regulate insulin release from β cells in the pancreatic islets as well as glucose uptake in the muscle cells together with αMSH and/or PRL. In addition, secreted CXCL14 with αMSH and/or PRL may act as a bioactive factor regulating hormone release in the adenohypophyseal cells of the reptilian pars distalis.
Sigma-Aldrich
Anti-Melanocyte Stimulating Hormone α Antibody, serum, Chemicon®
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey