Merck
  • Gut-Innervating Nociceptor Neurons Regulate Peyer's Patch Microfold Cells and SFB Levels to Mediate Salmonella Host Defense.
Gut-Innervating Nociceptor Neurons Regulate Peyer's Patch Microfold Cells and SFB Levels to Mediate Salmonella Host Defense.
Cell (2019-12-10)
Nicole Y Lai, Melissa A Musser, Felipe A Pinho-Ribeiro, Pankaj Baral, Amanda Jacobson, Pingchuan Ma, David E Potts, Zuojia Chen, Donggi Paik, Salima Soualhi, Yiqing Yan, Aditya Misra, Kaitlin Goldstein, Valentina N Lagomarsino, Anja Nordstrom, Kisha N Sivanathan, Antonia Wallrapp, Vijay K Kuchroo, Roni Nowarski, Michael N Starnbach, Hailian Shi, Neeraj K Surana, Dingding An, Chuan Wu, Jun R Huh, Meenakshi Rao, Isaac M Chiu
ABSTRACT
Gut-innervating nociceptor sensory neurons respond to noxious stimuli by initiating protective responses including pain and inflammation; however, their role in enteric infections is unclear. Here, we find that nociceptor neurons critically mediate host defense against the bacterial pathogen Salmonella enterica serovar Typhimurium (STm). Dorsal root ganglia nociceptors protect against STm colonization, invasion, and dissemination from the gut. Nociceptors regulate the density of microfold (M) cells in ileum Peyer's patch (PP) follicle-associated epithelia (FAE) to limit entry points for STm invasion. Downstream of M cells, nociceptors maintain levels of segmentous filamentous bacteria (SFB), a gut microbe residing on ileum villi and PP FAE that mediates resistance to STm infection. TRPV1+ nociceptors directly respond to STm by releasing calcitonin gene-related peptide (CGRP), a neuropeptide that modulates M cells and SFB levels to protect against Salmonella infection. These findings reveal a major role for nociceptor neurons in sensing and defending against enteric pathogens.
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
Anti-Neurofilament H Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide (CGRP) Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-Capsaicin Receptor Antibody, CT, serum, Chemicon®