Merck
Plasma lipid composition and risk of developing cardiovascular disease.
PloS one (2013-08-24)
Celine Fernandez, Marianne Sandin, Julio L Sampaio, Peter Almgren, Krzysztof Narkiewicz, Michal Hoffmann, Thomas Hedner, Björn Wahlstrand, Kai Simons, Andrej Shevchenko, Peter James, Olle Melander
ABSTRACT
We tested whether characteristic changes of the plasma lipidome in individuals with comparable total lipids level associate with future cardiovascular disease (CVD) outcome and whether 23 validated gene variants associated with coronary artery disease (CAD) affect CVD associated lipid species. Screening of the fasted plasma lipidome was performed by top-down shotgun analysis and lipidome compositions compared between incident CVD cases (n = 211) and controls (n = 216) from the prospective population-based MDC study using logistic regression adjusting for Framingham risk factors. Associations with incident CVD were seen for eight lipid species (0.21≤q≤0.23). Each standard deviation unit higher baseline levels of two lysophosphatidylcholine species (LPC), LPC16∶0 and LPC20∶4, was associated with a decreased risk for CVD (P = 0.024-0.028). Sphingomyelin (SM) 38∶2 was associated with increased odds of CVD (P = 0.057). Five triglyceride (TAG) species were associated with protection (P = 0.031-0.049). LPC16∶0 was negatively correlated with the carotid intima-media thickness (P = 0.010) and with HbA1c (P = 0.012) whereas SM38∶2 was positively correlated with LDL-cholesterol (P = 0.0*10(-6)) and the q-values were good (q≤0.03). The risk allele of 8 CAD-associated gene variants showed significant association with the plasma level of several lipid species. However, the q-values were high for many of the associations (0.015≤q≤0.75). Risk allele carriers of 3 CAD-loci had reduced level of LPC16∶0 and/or LPC 20∶4 (P≤0.056). Our study suggests that CVD development is preceded by reduced levels of LPC16∶0, LPC20∶4 and some specific TAG species and by increased levels of SM38∶2. It also indicates that certain lipid species are intermediate phenotypes between genetic susceptibility and overt CVD. But it is a preliminary study that awaits replication in a larger population because statistical significance was lost for the associations between lipid species and future cardiovascular events when correcting for multiple testing.
Product Description
Avanti
4ME 16:0 Diether PE, Avanti Research - A Croda Brand 999985P, powder
Pricing and availability is not currently available.