Merck
  • Structural Characterization of an Archaeal Lipid Bilayer as a Function of Hydration and Temperature.
Structural Characterization of an Archaeal Lipid Bilayer as a Function of Hydration and Temperature.
International journal of molecular sciences (2020-03-12)
Marta Salvador-Castell, Bruno Demé, Philippe Oger, Judith Peters
ABSTRACT
Archaea, the most extremophilic domain of life, contain ether and branched lipids which provide extraordinary bilayer properties. We determined the structural characteristics of diether archaeal-like phospholipids as functions of hydration and temperature by neutron diffraction. Hydration and temperature are both crucial parameters for the self-assembly and physicochemical properties of lipid bilayers. In this study, we detected non-lamellar phases of archaeal-like lipids at low hydration levels, and lamellar phases at levels of 90% relative humidity or more exclusively. Moreover, at 90% relative humidity, a phase transition between two lamellar phases was discernible. At full hydration, lamellar phases were present up to 70ᵒC and no phase transition was observed within the temperature range studied (from 25 °C to 70 °C). In addition, we determined the neutron scattering length density and the bilayer's structural parameters from different hydration and temperature conditions. At the highest levels of hydration, the system exhibited rearrangements on its corresponding hydrophobic region. Furthermore, the water uptake of the lipids examined was remarkably high. We discuss the effect of ether linkages and branched lipids on the exceptional characteristics of archaeal phospholipids.
Product Description
Avanti
4ME 16:0 Diether PE, Avanti Research - A Croda Brand 999985P, powder
Pricing and availability is not currently available.