Merck
  • DGKA Provides Platinum Resistance in Ovarian Cancer Through Activation of c-JUN-WEE1 Signaling.
DGKA Provides Platinum Resistance in Ovarian Cancer Through Activation of c-JUN-WEE1 Signaling.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-04-29)
Jie Li, Chaoyun Pan, Austin C Boese, JiHoon Kang, Anna D Umano, Kelly R Magliocca, Wenqing Yang, Yu Zhang, Sagar Lonial, Lingtao Jin, Sumin Kang
ABSTRACT
Although platinum compounds are the first-line treatment for ovarian cancer, the majority of patients relapse and develop resistance to treatment. However, the mechanism underlying resistance is unclear. The goal of our study is to decipher the mechanism by which a metabolic kinase, diacylglycerol kinase alpha (DGKA), confers platinum resistance in ovarian cancer. Metabolic kinase RNAi synthetic lethal screening was used to identify a cisplatin resistance driver in ovarian cancer. DGKA variants were used to demonstrate the need for DGKA activity in cisplatin resistance. Phospho-proteomic and genomic screens were performed to identify downstream effectors of DGKA. Therapeutic efficacy of targeting DGKA was confirmed and clinical relevance of DGKA signaling was validated using ovarian cancer patient-derived tumors that had different responses to platinum-based therapy. We found that platinum resistance was mediated by DGKA and its product, phosphatidic acid (PA), in ovarian cancer. Proteomic and genomic screens revealed that DGKA activates the transcription factor c-JUN and consequently enhances expression of a cell-cycle regulator, WEE1. Mechanistically, PA facilitates c-JUN N-terminal kinase recruitment to c-JUN and its nuclear localization, leading to c-JUN activation upon cisplatin exposure. Pharmacologic inhibition of DGKA sensitized ovarian cancer cells to cisplatin treatment and DGKA-c-JUN-WEE1 signaling positively correlated with platinum resistance in tumors derived from patients with ovarian cancer. Our study demonstrates how the DGKA-derived lipid messenger, PA, contributes to cisplatin resistance by intertwining with kinase and transcription networks, and provides preclinical evidence for targeting DGKA as a new strategy in ovarian cancer treatment to battle cisplatin resistance.
Sigma-Aldrich
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human DGKA
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIB3
Sigma-Aldrich
Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in mouse, clone GST-2, ascites fluid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Avanti
08:0 PA, 1,2-dioctanoyl-sn-glycero-3-phosphate (sodium salt), powder
Avanti
08:0 PA, 1,2-dioctanoyl-sn-glycero-3-phosphate (sodium salt), chloroform