Merck
  • EGCG protects vascular endothelial cells from oxidative stress-induced damage by targeting the autophagy-dependent PI3K-AKT-mTOR pathway.
EGCG protects vascular endothelial cells from oxidative stress-induced damage by targeting the autophagy-dependent PI3K-AKT-mTOR pathway.
Annals of translational medicine (2020-04-21)
Jiao Meng, Yuhua Chen, Junzhe Wang, Junling Qiu, Cuicui Chang, Fangfang Bi, Xiaopeng Wu, Wei Liu
ABSTRACT
Autophagy plays an important role in cellular homeostasis. Epigallocatechin gallate (EGCG), a polyphenol derived from green tea, has been shown to elicit vascular protective effects. Our study aimed to investigate the protective effect of EGCG in an endothelial injury model induced by hydrogen peroxide (H2O2) and reveal the possible mechanisms. Human vascular endothelial cells (HUVECs) were pretreatment with different concentration of EGCG, then exposed to H2O2. Cell viability was measured with MTS assay. Apoptosis was evaluated with TUNEL staining and apoptosis-related protein was determined by western blot. Autophagy flux was assessed by transmission electron microscopy and LC3 plasmid transfection. Besides, the role mTOR in EGCG-mediated antioxidant responses was validated with siRNA transfection. The results showed that pretreatment with EGCG significantly improved the survival of HUVECs from H2O2-induced cell death. After exposed to H2O2, EGCG upregulated the levels of Atg5, Atg7, LC3 II/I, and the Atg5-Atg12 complex in HUVECs, while downregulated apoptosis-related protein. Besides, EGCG inhibited the PI3K-AKT-mTOR signaling pathway. Knockdown of mTOR partially promoted EGCG-induced autophagy. These results suggest that EGCG induces autophagy by targeting the mTOR pathway, indicating that EGCG has the potential to prevent and treat oxidative stress-related cardiovascular diseases.
Product Description
Millipore
MCE Membrane Filter, 0.45 μm Pore Size, Immobilon®, filter diam. 90 mm, hydrophilic
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Anti-γ-Actin antibody, Mouse monoclonal, clone 2-2.1.14.17, purified from hybridoma cell culture