Merck
  • Micelles based on gold-glycopolymer complexes as new chemotherapy drug delivery agents.
Micelles based on gold-glycopolymer complexes as new chemotherapy drug delivery agents.
Chemical communications (Cambridge, England) (2012-04-05)
Samuel Pearson, Wei Scarano, Martina H Stenzel
ABSTRACT
Polymeric versions of deacetylated auranofin, a gold complex with a sugar ligand, were prepared by post-modifying RAFT glycopolymers. Micellisation of a block copolymer containing pendant Au(I) units produced nanoparticles with an increased anti-proliferative effect against OVCAR-3 human ovarian carcinoma cells.
Sigma-Aldrich
Auranofin, ≥98% (HPLC)