Merck
  • Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Developmental cell (2020-02-16)
Lisandra Vila Ellis, Margo P Cain, Vera Hutchison, Per Flodby, Edward D Crandall, Zea Borok, Bin Zhou, Edwin J Ostrin, Joshua D Wythe, Jichao Chen
ABSTRACT
The lung microvasculature is essential for gas exchange and commonly considered homogeneous. We show that VEGFA from the epithelium is required for a distinct endothelial cell (EC) population in the mouse lung. Vegfa is predominantly expressed by alveolar type 1 (AT1) cells and locally required to specify a subset of ECs. Single-cell RNA sequencing (scRNA-seq) reveals that ∼15% of lung ECs are transcriptionally distinct-marked by Carbonic anhydrase 4 (Car4)-and arise from bulk ECs, as suggested by trajectory analysis. Car4 ECs have extensive cellular projections and are separated from AT1 cells by a limited basement membrane without intervening pericytes. Car4 ECs are specifically lost upon epithelial Vegfa deletion; without Car4 ECs, the alveolar space is aberrantly enlarged despite the normal appearance of myofibroblasts. Lung Car4 ECs and retina tip ECs have common and distinct features. These findings support a signaling role of AT1 cells and shed light on alveologenesis.
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Fmoc-Cys(Trt) Wang resin, extent of labeling: ~0.7 mmol/g loading
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds