Merck
  • Assessment of Biodegradation Efficiency of Polychlorinated Biphenyls (PCBs) and Petroleum Hydrocarbons (TPH) in Soil Using Three Individual Bacterial Strains and Their Mixed Culture.
Assessment of Biodegradation Efficiency of Polychlorinated Biphenyls (PCBs) and Petroleum Hydrocarbons (TPH) in Soil Using Three Individual Bacterial Strains and Their Mixed Culture.
Molecules (Basel, Switzerland) (2020-02-12)
Teresa Steliga, Katarzyna Wojtowicz, Piotr Kapusta, Joanna Brzeszcz
ABSTRACT
Biodegradation is one of the most effective and profitable methods for the elimination of toxic polychlorinated biphenyls (PCBs) and total petroleum hydrocarbons (TPH) from the environment. In this study, aerobic degradation of the mentioned pollutants by bacterial strains Mycolicibacteriumfrederiksbergense IN53, Rhodococcuserythropolis IN129, and Rhodococcus sp. IN306 and mixed culture M1 developed based on those strains at 1:1:1 ratio was analyzed. The effectiveness of individual strains and of the mixed culture was assessed based on carried out respirometric tests and chromatographic analyses. The Rhodococcus sp. IN306 turned out most effective in terms of 18 PCB congeners biodegradation (54.4%). The biodegradation index was decreasing with an increasing number of chlorine atoms in a molecule. Instead, the Mycolicobacteriumfrederiksbergense IN53 was the best TPH degrader (37.2%). In a sterile soil, contaminated with PCBs and TPH, the highest biodegradation effectiveness was obtained using inoculation with mixed culture M1, which allowed to reduce both the PCBs (51.8%) and TPH (34.6%) content. The PCBs and TPH biodegradation capacity of the defined mixed culture M1 was verified ex-situ with prism method in a non-sterile soil polluted with aged petroleum hydrocarbons (TPH) and spent transformer oil (PCBs). After inoculation with mixed culture M1, the PCBs were reduced during 6 months by 84.5% and TPH by 70.8% as well as soil toxicity was decreased.
Product Description
Supelco
Phytane, analytical standard
Supelco
PCB Congeners in Soil, certified reference material
Supelco
Nitrate Test, photometric, 0.10-25.0 mg/L (NO3-N), 0.4-110.7 mg/L (NO3-), Spectroquant®
Supelco
Ammonium Test, photometric, 0.010-3.00 mg/L (NH4-N), Spectroquant®