Merck
  • Fusion of single proteoliposomes with planar, cushioned bilayers in microfluidic flow cells.
Fusion of single proteoliposomes with planar, cushioned bilayers in microfluidic flow cells.
Nature protocols (2012-04-21)
Erdem Karatekin, James E Rothman
ABSTRACT
Many biological processes rely on membrane fusion, and therefore assays to study its mechanisms are necessary. Here we report an assay with sensitivity to single-vesicle, and even to single-molecule events using fluorescently labeled vesicle-associated v-SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptor) liposomes and target-membrane-associated t-SNARE-reconstituted planar, supported bilayers (t-SBLs). Docking and fusion events can be detected using conventional far-field epifluorescence or total internal reflection fluorescence microscopy. In this assay, fusion is dependent on SNAP-25, one of the t-SNARE subunits that is required for fusion in vivo. The success of the assay is due to the use of: (i) bilayers covered with a thin layer of poly(ethylene glycol) (PEG) to control bilayer-bilayer and bilayer-substrate interactions, and (ii) microfluidic flow channels that present many advantages, such as the removal of nonspecifically bound liposomes by flow. The protocol takes 6-8 d to complete. Analysis can take up to 2 weeks.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Avanti
18:1 PEG2000 PE, Avanti Research - A Croda Brand 880130P, powder
Avanti
18:1 PEG2000 PE, Avanti Research - A Croda Brand 880130C
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 NBD PE, Avanti Research - A Croda Brand
Avanti
18:1 NBD PE, Avanti Research - A Croda Brand