Merck
  • High Open-Circuit Voltage Solar Cells Based on Organic-Inorganic Lead Bromide Perovskite.
High Open-Circuit Voltage Solar Cells Based on Organic-Inorganic Lead Bromide Perovskite.
The journal of physical chemistry letters (2013-03-21)
Eran Edri, Saar Kirmayer, David Cahen, Gary Hodes
ABSTRACT
Mesoscopic solar cells, based on solution-processed organic-inorganic perovskite absorbers, are a promising avenue for converting solar to electrical energy. We used solution-processed organic-inorganic lead halide perovskite absorbers, in conjunction with organic hole conductors, to form high voltage solar cells. There is a dire need for low-cost cells of this type, to drive electrochemical reactions or as the high photon energy cell in a system with spectral splitting. These perovskite materials, although spin-coated from solution, form highly crystalline materials. Their simple synthesis, along with high chemical versatility, allows tuning their electronic and optical properties. By judicious selection of the perovskite lead halide-based absorber, matching organic hole conductor, and contacts, a cell with a ∼ 1.3 V open circuit voltage was made. While further study is needed, this achievement provides a general guideline for additional improvement of cell performance.
Sigma-Aldrich
Di-n-propylammonium bromide
Sigma-Aldrich
Di-isopropylammonium bromide
Sigma-Aldrich
N,N-Dimethylethane-1,2-diammonium bromide
Sigma-Aldrich
Hexane-1,6-diammonium bromide
Sigma-Aldrich
N,N-Diethylpropane-1,3-diammonium bromide
Sigma-Aldrich
n-Dodecylammonium bromide
Sigma-Aldrich
N,N-Diethylethane-1,2-diammonium bromide
Sigma-Aldrich
n-Hexylammonium bromide
Sigma-Aldrich
n-Octylammonium bromide
Sigma-Aldrich
Butane-1,4-diammonium bromide
Sigma-Aldrich
Morpholinium bromide