Merck
  • Signaling transduction regulated by 5-hydroxytryptamine 1A receptor and orexin receptor 2 heterodimers.
Signaling transduction regulated by 5-hydroxytryptamine 1A receptor and orexin receptor 2 heterodimers.
Cellular signalling (2018-11-28)
Qin-Qin Wang, Chun-Mei Wang, Bao-Hua Cheng, Chun-Qing Yang, Bo Bai, Jing Chen
ABSTRACT
As G-protein-coupled receptors (GPCRs), 5-hydroxytryptamine 1A receptor (5-HT1AR) and orexin receptor 2 (OX2R) regulate the levels of the cellular downstream molecules. The heterodimers of different GPCRs play important roles in various of neurological diseases. Moreover, 5-HT1AR and OX2R are involved in the pathogenesis of neurological diseases such as depression with deficiency of hippocampus plasticity. However, the direct interaction of the two receptors remains elusive. In the present study, we firstly demonstrated the heterodimer formation of 5-HT1AR and OX2R. Exchange protein directly activated by cAMP (Epac) cAMP bioluminescence resonance energy transfer (BRET) biosensor analysis revealed that the expression levels of cellular cAMP significantly increased in HEK293T cells transfected with the two receptors compared with the 5-HT1AR group. Additionally, the cellular level of calcium was upregulated robustly in HEK293T cells co-transfected with 5-HT1AR and OX2R group after agonist treatment. Furthermore, western blotting data showed that 5-HT1AR and OX2R heterodimer decreased the levels of phosphorylation of extracellular signal-regulated kinase (ERK) and cAMP-response element-binding protein (CREB). These results not only unraveled the formation of 5-HT1AR and OX2R heterodimer but also suggested that the heterodimer affected the downstream signaling pathway, which will provide new insights into the function of the two receptors in the brain.
Sigma-Aldrich
Anti-Orexin-2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Htr1a antibody produced in goat, affinity isolated antibody, buffered aqueous solution