Merck
  • Characterization and kinetic analysis of the intracellular domain of human protein tyrosine phosphatase beta (HPTP beta) using synthetic phosphopeptides.
Characterization and kinetic analysis of the intracellular domain of human protein tyrosine phosphatase beta (HPTP beta) using synthetic phosphopeptides.
The Biochemical journal (1994-03-01)
K W Harder, P Owen, L K Wong, R Aebersold, I Clark-Lewis, F R Jirik
ABSTRACT
The intracellular domain of human protein tyrosine phosphatase beta (HPTP beta) (44 kDa) was expressed in bacteria, purified using epitope 'tagging' immunoaffinity chromatography, and characterized with respect to kinetic profile, substrate specificity and potential modulators of enzyme activity. A chromogenic assay based on the Malachite Green method was employed for the detection of inorganic phosphate (Pi) released from phosphopeptides by HPTP beta. This assay, modified so as to improve its sensitivity, was adapted to a 96-well microtitre plate format, and provided linear detection between 50 and 1000 pmol of Pi. The cytoplasmic domain of HPTP beta was strongly inhibited by vanadate, molybdate, heparin, poly(Glu, Tyr) (4:1) and zinc ions. In order to explore the substrate preferences of this PTPase, we generated 13-residue synthetic phosphotyrosine-containing peptides that corresponded to sites of physiological tyrosine phosphorylation. HPTP beta demonstrated kcat. values between 76 and 258 s-1 using four different phosphopeptides. The substrate preference of HPTP beta was in the order srcTyr-527 > PDGF-RTyr-740 > ERK1Tyr-204 > CSF-1RTyr-708 with Km values ranging from 140 microM to greater than 10 mM. The variations in affinity were probably due to differences among the four phosphopeptides compared, particularly with respect to the character of the charged amino acids flanking the phosphotyrosine residue.
Sigma-Aldrich
PTP Assay Kit 1, PTP Assay Kit 1 can be used to detect PTP-1B activity by dephosphorylation of the phosphopeptide (RRLIEDAEpYAARG) using malachite green detection, or by hydrolysis of pNPP.
Sigma-Aldrich
Tyrosine Phosphopeptide (RRLIEDAEpYAARG]
Sigma-Aldrich
Alkaline/Acid Phosphatase Assay Kit (R-R-A-pS-V-A), Alkaline/Acid Phosphatase Assay Kit is routinely used to detect phosphatase activity by either dephosphorylation of the phosphopeptide (RRApSVA) or hydrolysis of pNPP.
Sigma-Aldrich
Threonine Phosphopeptide (K-R-pT-I-R-R)
Sigma-Aldrich
Serine Phosphopeptide (RRApSVA)
Sigma-Aldrich
Ser/Thr Phosphatase Assay Kit 1 (K-R-pT-I-R-R), Ser/Thr Phosphatase Assay Kit 1used to detect PP2A activity by either dephosphorylation of the phosphopeptide.