Merck
  • Pteroyl-gamma-glutamate-cysteine synthesis and its application in folate receptor-mediated cancer cell targeting using folate-tethered liposomes.
Pteroyl-gamma-glutamate-cysteine synthesis and its application in folate receptor-mediated cancer cell targeting using folate-tethered liposomes.
Analytical biochemistry (2004-08-11)
Yonghua Zhang, Lili Guo, Roger W Roeske, Aśok C Antony, Hiremagalur N Jayaram
ABSTRACT
Cell membrane-associated folate receptors are selectively overexpressed in certain human tumors. The high affinity of folic acid for folate receptors provides a unique opportunity to use folic acid as a targeting ligand to deliver chemotherapeutic agents to cancer cells. Folate-tethered liposomes bearing pteroyl-gamma-glutamate-cysteine-polyethylene glycol (PEG)-distearoylphosphatidylethanolamine (DSPE) as the targeting component are under investigation as mediators of drug and gene delivery to cancer cells that overexpress folate receptors. Pteroyl-gamma-glutamate-cysteine synthesis is one of the crucial starting steps in the preparation of pteroyl-gamma-glutamate-cysteine-PEG-DSPE. However, published methods for the synthesis of pteroyl-gamma-glutamate-cysteine provide low yields and are not easily reproducible. Therefore, we developed a modified synthetic method for the removal of the N(10)-trifluoroacetyl group after cleavage/deprotection that is reliable, is easily reproducible, and has high yield (38%) compared with an unreliable yield of 3-20% with the earlier methods. Folate-tethered liposomes containing calcein or doxorubicin were prepared using pteroyl-gamma-glutamate-cysteine-PEG-DSPE as the targeting component along with nontargeted liposomes with PEG-DSPE. The results of the uptake of calcein and cytotoxicity of doxorubicin in human cervical cancer HeLa-IU(1) cells and human colon cancer Caco-2 cells demonstrated that folate-tethered liposomes were efficient in selective delivery to cancer cells overexpressing folate receptors. The improvement in yield of the targeting component can significantly facilitate "scale up" of folate receptor-mediated liposomal cancer therapy to the preclinical and clinical levels of investigations.
Product Description
Avanti
DSPE-PEG(2000) Folate, Avanti Research - A Croda Brand 880124P, powder
Avanti
DSPE-PEG(2000) Folate, Avanti Research - A Croda Brand 880124X
Avanti
DSPE-PEG(5000) Folate, Avanti Research - A Croda Brand 880123P, powder
Avanti
DSPE-PEG(5000) Folate, Avanti Research - A Croda Brand 880123X