Merck
  • Biodegradation of anthracene and several PAHs by the marine-derived fungus Cladosporium sp. CBMAI 1237.
Biodegradation of anthracene and several PAHs by the marine-derived fungus Cladosporium sp. CBMAI 1237.
Marine pollution bulletin (2017-10-23)
Willian G Birolli, Darlisson de A Santos, Natália Alvarenga, Anuska C F S Garcia, Luciane P C Romão, André L M Porto
ABSTRACT
The biodegradation of polycyclic aromatic hydrocarbons (PAHs) by marine-derived fungi was reported in this work. Marine-derived fungi (Trichoderma harzianum CBMAI 1677, Cladosporium sp. CBMAI 1237, Aspergillus sydowii CBMAI 935, Penicillium citrinum CBMAI 1186 and Mucor racemosus CBMAI 847) biodegraded anthracene (14days, 130rpm, 50mgmL-1 initial concentration in malt 2% medium). Cladosporium sp. CBMAI 1237 was the most efficient strain and biodegraded more anthracene in the presence (42% biodegradation) than in the absence (26%) of artificial seawater, suggesting that the biodegradation of PAHs may be faster in seawater than in non-saline environment. After 21days, Cladosporium sp. CBMAI 1237 biodegraded anthracene (71% biodegradation), anthrone (100%), anthraquinone (32%), acenaphthene (78%), fluorene (70%), phenanthrene (47%), fluoranthene (52%), pyrene (62%) and nitropyrene (64%). Previous undocumented metabolites were identified and, anthraquinone was a common product of different PAHs biodegradation. The marine-derived fungus Cladosporium sp. CBMAI 1237 showed potential for bioremediation of PAHs.
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Anthrone, ACS reagent, 97%
Sigma-Aldrich
Fluorene, 98%
Sigma-Aldrich
1-Nitropyrene, ≥95%
Sigma-Aldrich
Acenaphthene, 99%