Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zipper-top poly bag
Products
Product Category
bags (5)
Agency
Application
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z162949
Zipper-top poly bag,
size 3 in. × 5 in.
Expand
View Pricing
Z162957
Zipper-top poly bag,
size 4 in. × 6 in.
Expand
View Pricing
Z162965
Zipper-top poly bag,
size 5 in. × 8 in.
Expand
View Pricing
Z162973
Zipper-top poly bag,
size 9 in. × 12 in.
Expand
View Pricing
Z162930
Zipper-top poly bag,
size 2 1/2 in. × 3 in.
Expand
View Pricing