Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc sulfate heptahydrate
Linear Formula:
ZnSO
4
· 7H
2
O
CAS Number:
7446-20-0
Molecular Weight:
287.56
EC Number:
231-793-3
Products
Product Category
salts (10)
Brand
SAFC (2)
Sigma-Aldrich (2)
Supelco (1)
Sustainability
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
GMO free (1)
Halal (1)
Kosher (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
221376
Zinc sulfate heptahydrate,
ACS reagent, 99%
Expand
View Pricing
Z4750
Zinc sulfate heptahydrate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
31665
Zinc sulfate heptahydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Expand
View Pricing
Z0251
Zinc sulfate heptahydrate,
BioReagent, suitable for cell culture
Expand
View Pricing
1.08883
Zinc sulfate heptahydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
83265
Zinc sulfate heptahydrate,
BioUltra, Molecular Biology, 2.0 M in H
2
O
Expand
View Pricing
204986
Zinc sulfate heptahydrate,
≥99.95% trace metals basis
Expand
View Pricing
1.28209
Zinc sulfate heptahydrate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JP, USP
Expand
View Pricing
Z1001
Zinc sulfate heptahydrate,
suitable for plant cell culture
Expand
View Pricing
1.28210
Zinc sulfate heptahydrate,
FCC, GB
Expand
View Pricing
935662
Zinc sulfate heptahydrate,
99.999% trace metals basis
Expand
View Pricing
Z0635
Zinc sulfate heptahydrate,
meets USP testing specifications
Expand
View Pricing