Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Yttrium oxide
Products
Manufacturer
Physical Form
Diameter (mm)
Particle Size (µm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF06395676
Yttrium oxide,
powder, 5-10 μm mean particle size, weight 50 g, purity 99.9%
Expand
View Pricing
GF99459129
Yttrium oxide,
powder, 5-10 μm mean particle size, weight 100 g, purity 99.9%
Expand
View Pricing
GF32390456
Yttrium oxide,
powder, 5-10 μm mean particle size, weight 200 g, purity 99.9%
Expand
View Pricing
GF48807434
Yttrium oxide,
powder, 5-10 μm mean particle size, weight 500 g, purity 99.9%
Expand
View Pricing
GF09142613
Yttrium oxide,
sheet, thickness 2.0 mm, 10 mm diameter
Expand
View Pricing
GF34473146
Yttrium oxide,
sheet, thickness 0.5 mm, 10 mm diameter
Expand
View Pricing
GF53317515
Yttrium oxide,
sheet, thickness 6.0 mm, 10 mm diameter
Expand
View Pricing
GF42826541
Yttrium oxide,
sheet, thickness 3.0 mm, 10 mm diameter
Expand
View Pricing