Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Yttrium
Synonym(s):
Yttrium
Empirical Formula (Hill Notation):
Y
CAS Number:
7440-65-5
Molecular Weight:
88.91
Products
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Element
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
262994
Yttrium,
chips, 99.9% trace rare earth metals basis
Expand
View Pricing
GF87202009
Yttrium,
powder, 500 max. part. size (micron), weight 20 g, purity 99.9%
Expand
View Pricing
GF10436767
Yttrium,
powder, max. particle size 500 micron, weight 50 g, purity 99.9%
Expand
View Pricing
GF60321079
Yttrium,
powder, 500 max. part. size (micron), weight 100 g, purity 99.9%
Expand
View Pricing
GF55118257
Yttrium,
foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Expand
View Pricing
GF75315719
Yttrium,
foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Expand
View Pricing
GF68472692
Yttrium,
wire reel, 0.05m, diameter 0.5mm, 99.9%
Expand
View Pricing
GF40879008
Yttrium,
rod, 100mm, diameter 12.5mm, cast, 99%
Expand
View Pricing
GF92156711
Yttrium,
foil, thickness 0.25 mm, diameter 15 mm, purity 99%
Expand
View Pricing
GF84327037
Yttrium,
rod, 50mm, diameter 12.5mm, cast, 99%
Expand
View Pricing
GF32670772
Yttrium,
foil, thickness 0.1 mm, 12 mm diameter, purity 99%
Expand
View Pricing
GF11310275
Yttrium,
lump, 50 mm max. lump size, weight 50 g, purity 99.9%
Expand
View Pricing
GF96173017
Yttrium,
lump, 50 mm max. lump size, weight 20 g, purity 99.9%
Expand
View Pricing
GF35512725
Yttrium,
wire reel, 0.05m, diameter 1.0mm, 99.9%
Expand
View Pricing
GF12674617
Yttrium,
rod, 100mm, diameter 6.35mm, cast, 99%
Expand
View Pricing
GF68048860
Yttrium,
rod, 50mm, diameter 2.0mm, cast, 99%
Expand
View Pricing
GF11301398
Yttrium,
rod, 50mm, diameter 6.35mm, cast, 99%
Expand
View Pricing
GF02763963
Yttrium,
wire reel, 0.1m, diameter 0.5mm, 99.9%
Expand
View Pricing
GF85821701
Yttrium,
lump, 50 mm max. lump size, weight 100 g, purity 99.9%
Expand
View Pricing
GF86394156
Yttrium,
wire reel, 0.2m, diameter 1.0mm, 99.9%
Expand
View Pricing
Page 1 of 2
Page 1 of 2