Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xion AEM-Pention-72-15CL
Products
Brand
Sigma-Aldrich (15)
Sustainability
Feature
Physical Form
Length (cm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
AEM2B3044
Xion AEM-Pention-72-15CL,
thickness 30 μm, L × W 10 cm × 10 cm
Expand
View Pricing
AEM2B5066
Xion AEM-Pention-72-15CL,
thickness 50 μm, L × W 15 cm × 15 cm
Expand
View Pricing
AEM2B1022
Xion AEM-Pention-72-15CL,
thickness 10 μm, L × W 5 cm × 5 cm
Expand
View Pricing
AEM2B2044
Xion AEM-Pention-72-15CL,
thickness 20 μm, L × W 10 cm × 10 cm
Expand
View Pricing
AEM2B2066
Xion AEM-Pention-72-15CL,
thickness 20 μm, L × W 15 cm × 15 cm
Expand
View Pricing
AEM2B3022
Xion AEM-Pention-72-15CL,
thickness 30 μm, L × W 5 cm × 5 cm
Expand
View Pricing
AEM2B1044
Xion AEM-Pention-72-15CL,
thickness 10 μm, L × W 10 cm × 10 cm
Expand
View Pricing
AEM2B0566
Xion AEM-Pention-72-15CL,
thickness 5 μm, L × W 15 cm × 15 cm
Expand
View Pricing
AEM2B0522
Xion AEM-Pention-72-15CL,
thickness 5 μm, L × W 5 cm × 5 cm
Expand
View Pricing
AEM2B1066
Xion AEM-Pention-72-15CL,
thickness 10 μm, L × W 15 cm × 15 cm
Expand
View Pricing
AEM2B5044
Xion AEM-Pention-72-15CL,
thickness 50 μm, L × W 10 cm × 10 cm
Expand
View Pricing
AEM2B0544
Xion AEM-Pention-72-15CL,
thickness 5 μm, L × W 10 cm × 10 cm
Expand
View Pricing
AEM2B2022
Xion AEM-Pention-72-15CL,
thickness 20 μm, L × W 5 cm × 5 cm
Expand
View Pricing
AEM2B5022
Xion AEM-Pention-72-15CL,
thickness 50 μm, L × W 5 cm × 5 cm
Expand
View Pricing
AEM2B3066
Xion AEM-Pention-72-15CL,
thickness 30 μm, L × W 15 cm × 15 cm
Expand
View Pricing