Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wilmad
®
NMR tubes 10 mm diam., precision
Products
Product Category
tubes (11)
Brand
Sigma-Aldrich (6)
Manufacturer
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z274771
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 100 MHz
Expand
View Pricing
Z272051
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 500 MHz, L 7 in.
Expand
View Pricing
Z274720
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 400 MHz, L 7 in.
Expand
View Pricing
Z274755
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 500 MHz
Expand
View Pricing
Z274682
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 200 MHz, L 7 in.
Expand
View Pricing
Z274739
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 400 MHz, L 8 in.
Expand
View Pricing
Z272035
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 300 MHz, L 7 in.
Expand
View Pricing
Z274798
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 100 MHz
Expand
View Pricing
Z274747
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 400 MHz, L 9 in.
Expand
View Pricing
Z274801
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 100 MHz
Expand
View Pricing
Z274690
Wilmad
®
NMR tubes 10 mm diam., precision,
frequency 200 MHz, L 8 in.
Expand
View Pricing