Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray
Products
Product Category
vials (11)
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
DWK224882
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (4 mL), screw cap
Expand
View Pricing
DWK224884
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (8 mL), screw cap
Expand
View Pricing
DWK224881
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (2 mL), screw cap
Expand
View Pricing
DWK225288
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (20 mL), screw cap
Expand
View Pricing
DWK224885
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (12 mL), screw cap
Expand
View Pricing
DWK225289
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (25 mL), screw cap
Expand
View Pricing
DWK225012
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (4 mL), screw cap
Expand
View Pricing
DWK224888
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (24 mL), screw cap
Expand
View Pricing
DWK225290
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (40 mL), screw cap
Expand
View Pricing
DWK225014
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (8 mL), screw cap
Expand
View Pricing
DWK224886
WHEATON
®
clear sample vial with rubber lined cap packed in partitioned tray,
glass, tube capacity (16 mL), screw cap
Expand
View Pricing