Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Weigh Boat
Products
Product Category
dishes (4)
Color
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Size
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
HS1420BB
Weigh Boat,
Medium, square white polystyrene, Anti-Static, pack of 500 ea
Expand
View Pricing
HS1420AA
Weigh Boat,
size small, square white polystyrene, anti-static
Expand
View Pricing
HS1420CC
Weigh Boat,
Large, square white polystyrene, Anti-Static, pack of 500 ea
Expand
View Pricing
HS120223
Weigh Boat,
Medium, square blue polystyrene, Anti-static, pk of 500 ea
Expand
View Pricing
HS1420B
Weigh Boat,
Medium, square white polystyrene, pack of 500 ea
Expand
View Pricing
HS120710
Weigh Boat,
size medium, square white polystyrene, sterile; electron beam irradiated
Expand
View Pricing
HS1420C
Weigh Boat,
Large, square white polystyrene, pack of 500 ea
Expand
View Pricing
HS120222
Weigh Boat,
Small, square blue polystyrene, Anti-static, pk of 500 ea
Expand
View Pricing
HS1421B
Weigh Boat,
Medium, square blue polystyrene, pk of 500 ea
Expand
View Pricing
HS1421C
Weigh Boat,
Large, square blue polystyrene, pk of 500 ea
Expand
View Pricing
HS1421A
Weigh Boat,
Small, square blue polystyrene, pk of 500 ea
Expand
View Pricing
HS1420A
Weigh Boat,
size small, square white polystyrene
Expand
View Pricing
HS1423BB
Weigh Boat,
Medium, black polystyrene (square), Anti-static, pk of 500 ea
Expand
View Pricing
HS120880
Weigh Boat,
size small, square white polystyrene, anti-static, capacity 10 mL, sterile; electron beam irradiated
Expand
View Pricing
HS120224
Weigh Boat,
Large, square blue polystyrene, Anti-static, pk of 500 ea
Expand
View Pricing
HS1423AA
Weigh Boat,
Small, square black polystyrene, Anti-static, pk of 500 ea
Expand
View Pricing
HS1423C
Weigh Boat,
Large, square black polystyrene, pk of 500 ea
Expand
View Pricing
HS1423CC
Weigh Boat,
Large, square black polystyrene, Anti-static, pk of 500 ea
Expand
View Pricing
HS1423A
Weigh Boat,
Small, square black polystyrene, pk of 500 ea
Expand
View Pricing
HS1423B
Weigh Boat,
Medium, square black polystyrene, pk of 500 ea
Expand
View Pricing