Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw-top V-Vials
®
with solid top cap
Products
Product Category
vials (5)
Brand
Sigma-Aldrich (3)
Supelco (2)
Diameter (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z115118
Vials, screw-top V-Vials
®
with solid top cap,
volume 5 mL, thread for 20-400, diam. × H 20 mm × 65 mm, pkg of 12 ea
Expand
View Pricing
Z115061
Vials, screw-top V-Vials
®
with solid top cap,
volume 1 mL, thread for 13-425, diam. × H 13 mm × 44 mm, pkg of 12 ea
Expand
View Pricing
Z115088
Vials, screw-top V-Vials
®
with solid top cap,
volume 2 mL, thread for 15-415, diam. × H 17 mm × 61 mm, pkg of 12 ea
Expand
View Pricing
Z115096
Vials, screw-top V-Vials
®
with solid top cap,
volume 3 mL, thread for 20-400, diam. × H 20 mm × 50 mm
Expand
View Pricing
Z115053
Vials, screw-top V-Vials
®
with solid top cap,
volume 0.3 mL, thread for 13-425, diam. × H 13 mm × 35 mm, pkg of 12 ea
Expand
View Pricing