Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw top, graduated, open-top cap, preassembled
Products
Product Category
vials (7)
Brand
Supelco (6)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
27520-U
Vials, screw top, graduated, open-top cap, preassembled,
volume 22 mL, clear glass vial, thread for 20-400, O.D. × H 23 mm × 85 mm, PTFE/silicone septum
Expand
View Pricing
27521
Vials, screw top, graduated, open-top cap, preassembled,
volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, thread for 24-400, PTFE/silicone septum
Expand
View Pricing
27519
Vials, screw top, graduated, open-top cap, preassembled,
volume 15 mL, clear glass vial, thread for 18-400, O.D. × H 21 mm × 70 mm, PTFE/silicone septum
Expand
View Pricing
27518
Vials, screw top, graduated, open-top cap, preassembled,
volume 7 mL, clear glass vial, thread for 15-425, O.D. × H 17 mm × 60 mm, PTFE/silicone septum
Expand
View Pricing
27517
Vials, screw top, graduated, open-top cap, preassembled,
volume 4 mL, clear glass vial, thread for 13-425, O.D. × H 15 mm × 45 mm, PTFE/silicone septum
Expand
View Pricing
27526
Vials, screw top, graduated, open-top cap, preassembled,
volume 22 mL, clear glass vial, thread for 20-400, O.D. × H 23 mm × 85 mm
Expand
View Pricing
27527
Vials, screw top, graduated, open-top cap, preassembled,
volume 40 mL, clear glass vial, thread for 24-400, O.D. × H 29 mm × 82 mm, barrier liner (silicone bonded to aluminum)
Expand
View Pricing