Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tygon
®
formula 2375 laboratory tubing
Products
Product Category
tubing (13)
Manufacturer
ID (mm)
Length (m)
OD (mm)
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z685658
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 6.4 mm × 9.6 mm, L 15 m (50 ft)
Expand
View Pricing
Z685585
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 1.6 mm × 3.2 mm, L 15 m (50 ft)
Expand
View Pricing
Z685631
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 4.8 mm × 8.0 mm, L 15 m (50 ft)
Expand
View Pricing
Z685666
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 8.0 mm × 11.2 mm
Expand
View Pricing
Z685615
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 3.2 mm × 6.4 mm, L 15 m (50 ft)
Expand
View Pricing
Z685623
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 4.0 mm × 5.6 mm, L 15 m (50 ft)
Expand
View Pricing
Z685593
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 1.6 mm × 4.8 mm, L 15 m (50 ft)
Expand
View Pricing
Z685682
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 9.6 mm × 15.9 mm, L 15 m (50 ft)
Expand
View Pricing
Z685690
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 12.7 mm × 19.1 mm, L 15 m (50 ft)
Expand
View Pricing
Z685674
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 9.6 mm × 12.7 mm, L 15 m (50 ft)
Expand
View Pricing
Z685720
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 25.4 mm × 35.0 mm, L 7.5 m (25 ft)
Expand
View Pricing
Z685712
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 19.0 mm × 25.4 mm, L 15 m (50 ft)
Expand
View Pricing
Z685704
Tygon
®
formula 2375 laboratory tubing,
I.D. × O.D. 15.9 mm × 22.3 mm, L 15 m (50 ft)
Expand
View Pricing