Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten-rhenium alloy, W95Re5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF39259053
Tungsten-rhenium alloy, W95Re5,
0.5m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF13134169
Tungsten-rhenium alloy, W95Re5,
0.5m wire, 0.7mm diameter, annealed
Expand
View Pricing
GF34437988
Tungsten-rhenium alloy, W95Re5,
1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF66301264
Tungsten-rhenium alloy, W95Re5,
0.2m wire, 0.7mm diameter, annealed
Expand
View Pricing
GF18675502
Tungsten-rhenium alloy, W95Re5,
0.5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF20105323
Tungsten-rhenium alloy, W95Re5,
2m wire, 0.65mm diameter, annealed
Expand
View Pricing
GF88098250
Tungsten-rhenium alloy, W95Re5,
0.1m wire, 0.65mm diameter, annealed
Expand
View Pricing
GF99367172
Tungsten-rhenium alloy, W95Re5,
5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF58015953
Tungsten-rhenium alloy, W95Re5,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF39199517
Tungsten-rhenium alloy, W95Re5,
1m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF07382437
Tungsten-rhenium alloy, W95Re5,
1m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF18360929
Tungsten-rhenium alloy, W95Re5,
2m wire, 0.7mm diameter, annealed
Expand
View Pricing
GF68149323
Tungsten-rhenium alloy, W95Re5,
1m wire, 0.65mm diameter, annealed
Expand
View Pricing
GF44125736
Tungsten-rhenium alloy, W95Re5,
0.2m wire, 0.65mm diameter, annealed
Expand
View Pricing
GF04748261
Tungsten-rhenium alloy, W95Re5,
2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF12149401
Tungsten-rhenium alloy, W95Re5,
0.2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF20897196
Tungsten-rhenium alloy, W95Re5,
0.1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF62064121
Tungsten-rhenium alloy, W95Re5,
0.5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF10096512
Tungsten-rhenium alloy, W95Re5,
0.2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF65933444
Tungsten-rhenium alloy, W95Re5,
20m wire, 0.125mm diameter, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2