Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TSKgel
®
SP-5PW (20 µm) HPLC Columns
Synonym(s):
TSKgel
®
Cation Exchange HPLC Guard Kit
Products
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
808807
TSKgel
®
SP-5PW (20 µm) HPLC Columns,
L x I.D. 1 cm × 8 mm, sulfonic acid phase
Expand
View Pricing
807211
TSKgel
®
SP-5PW (20 µm) HPLC Columns,
L x I.D. 2.5 cm × 6 mm, sulfonic acid phase
Expand
View Pricing
816093
TSKgel
®
SP-5PW (20 µm) HPLC Columns,
L x I.D. 3.5 cm × 10 mm, sulfonic acid phase
Expand
View Pricing