![TSKgel® Boronate-5PW HPLC Column phase m-aminophenyl boronate, L × I.D. 5 cm × 5 mm, 10 μm particle size, glass](/deepweb/assets/sigmaaldrich/product/images/359/826/6fecccd2-7e32-4949-9ab0-39cb7adb7fa0/640/6fecccd2-7e32-4949-9ab0-39cb7adb7fa0.jpg)
TSKgel® Boronate-5PW (10 µm) HPLC Columns
![TSKgel® Boronate-5PW HPLC Column phase m-aminophenyl boronate, L × I.D. 5 cm × 5 mm, 10 μm particle size, glass](/deepweb/assets/sigmaaldrich/product/images/359/826/6fecccd2-7e32-4949-9ab0-39cb7adb7fa0/640/6fecccd2-7e32-4949-9ab0-39cb7adb7fa0.jpg)
Synonym(s):
TSKgel® Boronate-5PW HPLC Column
Products
Product Category
Manufacturer
Product Line
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2