Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypsin from porcine pancreas
CAS Number:
9002-07-7
EC Number:
232-650-8
Products
Product Category
proteases (4)
Brand
Sigma-Aldrich (6)
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
T4799
Trypsin from porcine pancreas,
lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Expand
View Pricing
T0303
Trypsin from porcine pancreas,
Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Expand
View Pricing
T7409
Trypsin from porcine pancreas,
lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Expand
View Pricing
T5266
Trypsin from porcine pancreas,
lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Expand
View Pricing
T7168
Trypsin from porcine pancreas,
tablet, 1 mg tablet
Expand
View Pricing
93615
Trypsin from porcine pancreas,
~1500 U/mg
Expand
View Pricing