Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TrueNorth
®
Mini Cooler
Products
Height (mm)
Wells
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
HS120074
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit -20 °C, size 32 wells, blue polycarbonate
Expand
View Pricing
HS120073
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit -20 °C, size 12 wells, blue polycarbonate
Expand
View Pricing
HS120075
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with gel-filled lid, temp. limit -20 °C, size 32 wells, blue polycarbonate
Expand
View Pricing
HS120071
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit 0 °C, size 32 wells, green polycarbonate
Expand
View Pricing
HS120070
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit 0 °C, size 12 wells, green polycarbonate
Expand
View Pricing
HS120076
TrueNorth
®
Mini Cooler,
for tubes, 0.2 mL and assay plates, with clear lid, temp. limit -20 °C, size 96 wells, blue polycarbonate
Expand
View Pricing
HS120072
TrueNorth
®
Mini Cooler,
for tubes, 0.5 ‑ 2 mL, with gel-filled lid, temp. limit 0 °C, size 32 wells, green polycarbonate
Expand
View Pricing