Skip to Content
Merck
Tris(triphenylphosphine)rhodium(I) chloride

Tris(triphenylphosphine)rhodium(I) chloride

Tris(triphenylphosphine)rhodium(I) chloride
Synonym(s):
NSC 124140, RhCl(PPh3)3, Rhodium(I) tris(triphenylphosphine) chloride, Wilkinson’s catalyst
Linear Formula:
[(C6H5)3P]3RhCl
CAS Number:
Molecular Weight:
925.22
EC Number:

Products

Product Category
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2