Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trioctylphosphine oxide
Synonym(s):
(Oct)
3
PO, TOPO
®
Linear Formula:
[CH
3
(CH
2
)
7
]
3
PO
CAS Number:
78-50-2
Molecular Weight:
386.63
EC Number:
201-121-3
Products
Product Category
reagents (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technical (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
223301
Trioctylphosphine oxide,
ReagentPlus
®
, 99%
Expand
View Pricing
346187
Trioctylphosphine oxide,
technical grade, 90%
Expand
View Pricing
00676
Trioctylphosphine oxide,
for extraction analysis, ≥98.5%
Expand
View Pricing