Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trimethyl borate
Synonym(s):
Boric acid trimethyl ester, Methyl borate, Trimethyl borate
Linear Formula:
B(OCH
3
)
3
CAS Number:
121-43-7
Molecular Weight:
103.91
EC Number:
204-468-9
Products
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Purum (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
452920
Trimethyl borate,
≥98%
Expand
View Pricing
443999
Trimethyl borate,
purified by redistillation, ≥99.5%
Expand
View Pricing
447218
Trimethyl borate,
99.999% (trace metal basis)
Expand
View Pricing
8.21180
Trimethyl borate,
for synthesis
Expand
View Pricing
92330
Trimethyl borate,
purum, ≥99.0% (GC)
Expand
View Pricing