Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TOYOPEARL
®
Phenyl-650C Bulk Media
Synonym(s):
TOYOPEARL
®
Phenyl-650 Bulk Media
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
814785
TOYOPEARL
®
Phenyl-650C Bulk Media,
phase phenyl, bottle of 1 L, 100 μm particle size
Expand
View Pricing
843126
TOYOPEARL
®
Phenyl-650C Bulk Media,
phase phenyl, bottle of 25 mL, 100 μm particle size
Expand
View Pricing