Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Total Base Number Standard
Synonym(s):
TBN-Standard
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
TBN10
Total Base Number Standard,
ASTM D2896 (10.01 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TBN70
Total Base Number Standard,
ASTM D2896 (70.24 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TBN6
Total Base Number Standard,
ASTM D2896 (6.06 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TBN15
Total Base Number Standard,
ASTM D2896 (15.17 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TBN30
Total Base Number Standard,
ASTM D2896 (30.17 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TBN1
Total Base Number Standard,
ASTM D 2896 (0.99 mg KOH/g), pkg of 1 × 125 g
Expand
View Pricing
TBN3
Total Base Number Standard,
ASTM D 2896 (3.01 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing