
Toluene-4-sulfonic acid monohydrate

Synonym(s):
Toluene-4-sulfonic acid monohydrate
Empirical Formula (Hill Notation):
C7H8O3S · xH2O
CAS Number:
Molecular Weight:
172.20 (anhydrous basis)
Products
Brand
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing