Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TOC standard
Products
Brand
Supelco (6)
Agency
Format
Quality Segment
Application
Special Grade
analytical standard (6)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
76067
TOC standard,
according to ISO/CEN EN1484, 1000 mg/L±10 mg/L
Expand
View Pricing
70961
TOC standard,
according to ISO/CEN EN1484, 100 mg/L±5 mg/L
Expand
View Pricing
90326
TOC standard,
25 mg/L±2 mg/L
Expand
View Pricing
76319
TOC standard,
according to ISO/CEN EN1484, 500 mg/L±10 mg/L
Expand
View Pricing
94450
TOC standard,
according to ISO/CEN EN1484, 50 mg/L±2 mg/L
Expand
View Pricing
95781
TOC standard,
according to ISO/CEN EN1484, 250 mg/L±5 mg/L
Expand
View Pricing