Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium/Aluminum/Vanadium
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Particle Size (µm)
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF83413694
Titanium/Aluminum/Vanadium,
foil, thickness 0.25 mm, Ti94.5%/Al 3%/V 2.5%, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF07785692
Titanium/Aluminum/Vanadium,
powder, Ti 90%/Al 6%/V 4%, max. particle size 200 micron, weight 50 g, min. particle size 150 micron
Expand
View Pricing
GF40862606
Titanium/Aluminum/Vanadium,
rod, Ti 90%/Al 6%/V 4%, 1.0 mm diameter, length 500 mm, temper annealed
Expand
View Pricing
GF52236064
Titanium/Aluminum/Vanadium,
rod, Ti 90%/Al 6%/V 4%, 1.0 mm diameter, length 100 mm, temper annealed
Expand
View Pricing
GF12641701
Titanium/Aluminum/Vanadium,
powder, Ti 90%/Al 6%/V 4%, max. particle size 200 micron, weight 100 g, min. particle size 150 micron
Expand
View Pricing
GF53787134
Titanium/Aluminum/Vanadium,
rod, Ti 90%/Al 6%/V 4%, 1.0 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF79014461
Titanium/Aluminum/Vanadium,
rod, Ti90%/Al 6%/V 4%, 1.0 mm diameter, length 1000 mm, temper annealed
Expand
View Pricing
GF11606419
Titanium/Aluminum/Vanadium,
foil, Ti 94.5%/Al 3%/V 2.5%, thickness 0.25 mm, length 0.5 m, temper annealed
Expand
View Pricing
GF12600762
Titanium/Aluminum/Vanadium,
rod, Ti 90%/Al 6%/V 4%, 2.0 mm diameter, length 500 mm, temper annealed
Expand
View Pricing
GF50839218
Titanium/Aluminum/Vanadium,
rod, Ti 90%/Al 6%/V 4%, 2.0 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF07632637
Titanium/Aluminum/Vanadium,
foil, Ti 94.5%/Al 3%/V 2.5%, thickness 0.25 mm, size 50 × 50 mm, temper annealed
Expand
View Pricing
GF23358078
Titanium/Aluminum/Vanadium,
powder, Ti 90%/Al 6%/V 4%, max. particle size 200 micron, weight 200 g, min. particle size 150 micron
Expand
View Pricing
GF47529647
Titanium/Aluminum/Vanadium,
foil, Ti 90%/Al 6%/V 4%, thickness 0.1 mm, length 0.5 m, coil width 12 mm
Expand
View Pricing
GF07295654
Titanium/Aluminum/Vanadium,
foil, Ti 90%/Al 6%/V 4%, thickness 0.1 mm, length 0.1 m, coil width 220 mm
Expand
View Pricing
GF48944043
Titanium/Aluminum/Vanadium,
foil, Ti 90%/Al 6%/V 4%, thickness 0.05 mm, length 0.2 m, coil width 120 mm
Expand
View Pricing
GF69352061
Titanium/Aluminum/Vanadium,
foil, Ti90%/Al 6%/V 4%, thickness 0.1 mm, length 0.2 m, coil width 220 mm
Expand
View Pricing
GF10946314
Titanium/Aluminum/Vanadium,
foil, Ti 90%/Al 6%/V 4%, thickness 0.05 mm, length 0.5 m, coil width 120 mm
Expand
View Pricing
GF26031002
Titanium/Aluminum/Vanadium,
foil, Ti 94.5%/Al 3%/V 2.5%, thickness 0.25 mm, size 150 × 150 mm, temper annealed
Expand
View Pricing
GF95578611
Titanium/Aluminum/Vanadium,
foil, Ti90%/Al 6%/V 4%, thickness 0.05 mm, length 0.1 m, coil width 120 mm
Expand
View Pricing
GF98836206
Titanium/Aluminum/Vanadium,
foil, thickness 0.8 mm, Ti90%/Al 6%/V 4%, size 50 × 50 mm, temper annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2