Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thrombin from bovine plasma
Synonym(s):
Factor IIa
CAS Number:
9002-04-4
EC Number:
232-648-7
Products
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T4648
Thrombin from bovine plasma,
lyophilized powder, 40-300 NIH units/mg protein (biuret)
Expand
View Pricing
T6634
Thrombin from bovine plasma,
lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
Expand
View Pricing
T7326
Thrombin from bovine plasma,
≥60 NIH units/mg protein (biuret)
Expand
View Pricing
T7513
Thrombin from bovine plasma,
lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
Expand
View Pricing
SRE0003
Thrombin from bovine plasma,
Suitable for manufacturing of diagnostic kits and reagents
Expand
View Pricing