Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thermogreen
®
LB-2 Septa, solid discs
Products
Product Category
septa (16)
Brand
Sigma-Aldrich (2)
Supelco (14)
Diameter (mm)
Compatibility
Quality Segment
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
20654
Thermogreen
®
LB-2 Septa, solid discs,
diam. 11 mm ( 7/16 in.), pkg of 50 ea
Expand
View Pricing
23163
Thermogreen
®
LB-2 Septa, solid discs,
diam. 11 mm ( 7/16), pkg of 250 ea
Expand
View Pricing
20652
Thermogreen
®
LB-2 Septa, solid discs,
diam. 9.5 mm ( 3/8 in.), pkg of 50 ea
Expand
View Pricing
23164
Thermogreen
®
LB-2 Septa, solid discs,
diam. 11 mm ( 7/16), pkg of 1000 ea
Expand
View Pricing
20638
Thermogreen
®
LB-2 Septa, solid discs,
diam. 5 mm ( 3/16 in.), pkg of 50 ea
Expand
View Pricing
23154
Thermogreen
®
LB-2 Septa, solid discs,
diam. 11.5 mm ( 11/24 in.), pkg of 50 ea
Expand
View Pricing
23159
Thermogreen
®
LB-2 Septa, solid discs,
diam. 17 mm ( 21/32 in.), pkg of 50 ea
Expand
View Pricing
20651
Thermogreen
®
LB-2 Septa, solid discs,
diam. 6 mm ( 1/4 in.), pkg of 50 ea
Expand
View Pricing
20660-U
Thermogreen
®
LB-2 Septa, solid discs,
diam. 12.5 mm ( 1/2 in.), pkg of 50 ea
Expand
View Pricing
20653-U
Thermogreen
®
LB-2 Septa, solid discs,
diam. 10 mm ( 13/32 in.), pkg of 50 ea
Expand
View Pricing
20666
Thermogreen
®
LB-2 Septa, solid discs,
diam. 9.5 mm ( 3/8 in.), pkg of 250 ea
Expand
View Pricing
28021-U
Thermogreen
®
LB-2 Septa, solid discs,
diam. 9 mm ( 11/32 in.), pkg of 50 ea
Expand
View Pricing
20663
Thermogreen
®
LB-2 Septa, solid discs,
diam. 16 mm ( 5/8 in.), pkg of 50 ea
Expand
View Pricing
23157
Thermogreen
®
LB-2 Septa, solid discs,
diam. 10 mm ( 13/32), pkg of 1000 ea
Expand
View Pricing
23156
Thermogreen
®
LB-2 Septa, solid discs,
diam. 10 mm ( 13/32 in.), pkg of 250 ea
Expand
View Pricing
20678
Thermogreen
®
LB-2 Septa, solid discs,
diam. 12.5 mm ( 1/2 in.), pkg of 250 ea
Expand
View Pricing