Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetramethylsilane
Synonym(s):
Tetramethylsilane
Linear Formula:
Si(CH
3
)
4
CAS Number:
75-76-3
Molecular Weight:
88.22
EC Number:
200-899-1
Products
Product Category
solvents (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
ACS reagent (1)
Puriss (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T24007
Tetramethylsilane,
ACS reagent, NMR grade, ≥99.9%
Expand
View Pricing
87921
Tetramethylsilane,
≥99.0% (GC)
Expand
View Pricing
1.08183
Tetramethylsilane,
for the calibration of NMR spectra for NMR spectroscopy MagniSolv
™
Expand
View Pricing
87920
Tetramethylsilane,
analytical standard, for NMR spectroscopy, ACS reagent
Expand
View Pricing
523771
Tetramethylsilane,
electronic grade, ≥99.99% trace metals basis
Expand
View Pricing