Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetraethylammonium hydroxide solution
Synonym(s):
TEA hydroxide
Linear Formula:
(C
2
H
5
)
4
N(OH)
CAS Number:
77-98-5
Molecular Weight:
147.26
Products
Product Category
organic bases (3)
Formula Weight
Physical Form
Quality Segment
Functional Group
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
302929
Tetraethylammonium hydroxide solution,
35 wt. % in H
2
O
Expand
View Pricing
177806
Tetraethylammonium hydroxide solution,
20 wt. % in H
2
O
Expand
View Pricing
86631
Tetraethylammonium hydroxide solution,
~25% in methanol (~1.5 M)
Expand
View Pricing
86636
Tetraethylammonium hydroxide solution,
~1.0 M (CH
3
CH
2
)
4
NOH in H
2
O, electrochemical grade
Expand
View Pricing