Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
reaction tube with high vacuum valve
Products
Product Category
tubes (6)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SYNF561025
Synthware
™
reaction tube with high vacuum valve,
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing
SYNF564100
Synthware
™
reaction tube with high vacuum valve,
100 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF564250
Synthware
™
reaction tube with high vacuum valve,
250 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF561100
Synthware
™
reaction tube with high vacuum valve,
100 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing
SYNF561050
Synthware
™
reaction tube with high vacuum valve,
50 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing
SYNF564350
Synthware
™
reaction tube with high vacuum valve,
350 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing