Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
pear-shaped Schlenk flask with stopcock
Products
Product Category
flasks (33)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
SYNF902450
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 50 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF901410
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 10 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF904500G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF904000
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 1,000 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF904200
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 200 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF901450
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF904000G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 1,000 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF901950
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 50 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF909100
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF901425
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF901450G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF901410G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 10 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF904100H
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF902450G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 50 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF904100
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF909100G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF901100
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF909200
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 200 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF904100G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF904200G
Synthware
™
pear-shaped Schlenk flask with stopcock,
capacity 200 mL, joint: ST/NS 24/40
Expand
View Pricing
Page 1 of 2
Page 1 of 2